Order Entry
Netherlands
ContactUsLinkComponent
 
Pentafluorophenyl 1-(thien-2-ylmethyl)piperidine-4-carboxylate 97%
  APOSPC3225-250MG
 :  
Pentafluorophenyl 1-(thien-2-ylmethyl)piperidine-4-carboxylate 97%
  APOSPC3225-250MG
 :  

 

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

 

 

 :
 :391.356 g/mol
 :73…75 °C
 :Refrigerator
 :2928
 :6.1+8,II
 :930111-06-1
 :C₁₇H₁₄F₅NO₂S
 :MFCD09879954