Order Entry
Netherlands
ContactUsLinkComponent
4-[3-(Trifluoromethyl)-1H-pyrazol-1-yl]benzoic acid
Irritant
4-[3-(Trifluoromethyl)-1H-pyrazol-1-yl]benzoic acid
Catalog # APOSPC31875-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-[3-(Trifluoromethyl)-1H-pyrazol-1-yl]benzoic acid
Catalog # APOSPC31875-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :256.181 g/mol
Melting Pt:189…190 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:220462-27-1
Formula:C₁₁H₇F₃N₂O₂
MDL Number:MFCD03086203