4-Chloro-6-(trifluoromethoxy)-2-(trifluoromethyl)quinoline
Catalog # APOSPC31863-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:315.599 g/mol
Storage Temperature:Ambient
CAS Number:503148-24-1
Formula:C₁₁H₄ClF₆NO
MDL Number:MFCD01566524