
Ethyl 4-chloro-2-(trifluoromethyl)quinoline-6-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:680211-86-3
:
Formula:C₁₃H₉ClF₃NO₂
MDL Number:MFCD00277503
:303.664 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together