
Methyl 3-chloro-5-(trifluoromethyl)pyridine-2-carboxylate
Catalog # APOSPC31856-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:239.579 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:655235-65-7
Formula:C₈H₅ClF₃NO₂
MDL Number:MFCD01570523