Order Entry
Netherlands
ContactUsLinkComponent
3-Bromo-4-methyl-5-(trifluoromethyl)benzenesulphonyl chloride
3-Bromo-4-methyl-5-(trifluoromethyl)benzenesulphonyl chloride
Catalog # APOSPC303610-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Bromo-4-methyl-5-(trifluoromethyl)benzenesulphonyl chloride
Catalog # APOSPC303610-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :337.541 g/mol
Storage Temperature:Ambient
CAS Number:2383371-10-4
Formula:C₈H₅BrClF₃O₂S
MDL Number:MFCD32640182