3-Bromo-4-methyl-5-(trifluoromethyl)phenylacetonitrile 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2384884-96-0
:
Formula:C₁₀H₇BrF₃N
MDL Number:MFCD32640175
:278.068 g/mol
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
Frequently Bought Together