Order Entry
Netherlands
ContactUsLinkComponent
3-Bromo-2-chloro-5-(trifluoromethyl)benzoic acid 97%
3-Bromo-2-chloro-5-(trifluoromethyl)benzoic acid 97%
Catalog # APOSPC303581-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Bromo-2-chloro-5-(trifluoromethyl)benzoic acid 97%
Catalog # APOSPC303581-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :303.46 g/mol
Storage Temperature:Ambient
CAS Number:2091224-21-2
Formula:C₈H₃BrClF₃O₂
MDL Number:MFCD32640158