3-Bromo-2-chloro-5-(trifluoromethyl)phenylacetonitrile
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2090294-87-2
:
Formula:C₉H₄BrClF₃N
MDL Number:MFCD32640155
:298.487 g/mol
Storage Temperature:Ambient
Frequently Bought Together