5-Bromo-3-chloro-2-fluorobenzoyl chloride
Catalog # APOSPC303458-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:271.899 g/mol
Storage Temperature:Ambient
CAS Number:1536225-35-0
Formula:C₇H₂BrCl₂FO
MDL Number:MFCD26686459