Order Entry
Netherlands
ContactUsLinkComponent
4-Chloro-3-(trifluoromethylthio)phenylacetonitrile 97%
4-Chloro-3-(trifluoromethylthio)phenylacetonitrile 97%
Catalog # APOSPC303392-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Chloro-3-(trifluoromethylthio)phenylacetonitrile 97%
Catalog # APOSPC303392-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :251.656 g/mol
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
CAS Number:2514625-02-4
Formula:C₉H₅ClF₃NS
MDL Number:MFCD32640053