2-Fluoro-3-methyl-5-(trifluoromethyl)benzyl bromide 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2091670-09-4
:
Formula:C₉H₇BrF₄
MDL Number:MFCD31916396
:271.049 g/mol
Storage Temperature:Ambient
Frequently Bought Together