3-Chloro-2-fluoro-4-(trifluoromethyl)aniline 98%
Catalog # APOSPC303375-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:213.56 g/mol
Storage Temperature:Ambient
UN:2810
ADR:6.1,III
CAS Number:1807135-05-2
Formula:C₇H₄ClF₄N
MDL Number:MFCD28752559