2-Fluoro-4-methyl-3-(trifluoromethyl)phenylacetonitrile 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1823323-72-3
:
Formula:C₁₀H₇F₄N
MDL Number:MFCD28103371
:217.163 g/mol
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
Frequently Bought Together