4-Fluoro-3-methyl-2-(trifluoromethyl)phenylacetonitrile 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1706446-60-7
:
Formula:C₁₀H₇F₄N
MDL Number:MFCD28054203
:217.163 g/mol
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
Frequently Bought Together