4-Methoxy-3-methyl-5-(trifluoromethyl)phenylacetonitrile 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1706430-20-7
:
Formula:C₁₁H₁₀F₃NO
MDL Number:MFCD27986834
:229.198 g/mol
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
Frequently Bought Together