Order Entry
Netherlands
ContactUsLinkComponent
3-Methyl-5-(trifluoromethoxy)phenylacetonitrile 98%
3-Methyl-5-(trifluoromethoxy)phenylacetonitrile 98%
Catalog # APOSPC302965-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Methyl-5-(trifluoromethoxy)phenylacetonitrile 98%
Catalog # APOSPC302965-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :215.172 g/mol
Storage Temperature:Ambient
UN:3276
ADR:6.1,III
CAS Number:916420-56-9
Formula:C₁₀H₈F₃NO
MDL Number:MFCD09025374