4-Fluoro-3-(trifluoromethoxy)phenylacetonitrile 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1020718-23-3
:
Formula:C₉H₅F₄NO
MDL Number:MFCD07772785
:219.136 g/mol
Boiling Pt:91 °C
Storage Temperature:Ambient
UN:3276
ADR:6.1,III
Frequently Bought Together