Order Entry
Netherlands
ContactUsLinkComponent
3-Chloro-2-fluoro-4-(trifluoromethyl)phenylacetonitrile 97%
3-Chloro-2-fluoro-4-(trifluoromethyl)phenylacetonitrile 97%
Catalog # APOSPC302711-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Chloro-2-fluoro-4-(trifluoromethyl)phenylacetonitrile 97%
Catalog # APOSPC302711-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :237.581 g/mol
Storage Temperature:Ambient
CAS Number:1431329-60-0
Formula:C₉H₄ClF₄N
MDL Number:MFCD23795330