
3-[3-Methyl-5-(trifluoromethyl)phenyl]propionic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1092460-65-5
:
Formula:C₁₁H₁₁F₃O₂
MDL Number:MFCD11519313
:232.199 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together