
2-(Difluoromethoxy)-5-fluorobenzoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1214383-11-5
:1017779-44-0
Formula:C₈H₅F₃O₃
MDL Number:MFCD09832364
:206.119 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together