Order Entry
Netherlands
ContactUsLinkComponent
N-[2-Methyl-6-(trifluoromethyl)pyridin-3-yl]-N'-pyridin-3-ylurea
N-[2-Methyl-6-(trifluoromethyl)pyridin-3-yl]-N'-pyridin-3-ylurea
Catalog # APOSPC300949-1G
Supplier:  Apollo Scientific
CAS Number:  
N-[2-Methyl-6-(trifluoromethyl)pyridin-3-yl]-N'-pyridin-3-ylurea
Catalog # APOSPC300949-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :296.248 g/mol
Storage Temperature:Ambient
Formula:C₁₃H₁₁F₃N₄O
MDL Number:MFCD29918614