N-[2-Methyl-6-(trifluoromethyl)pyridin-3-yl]morpholine-4-carboxamide
Catalog # APOSPC300927-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:289.254 g/mol
Storage Temperature:Ambient
Formula:C₁₂H₁₄F₃N₃O₂
MDL Number:MFCD30481167