Order Entry
Netherlands
ContactUsLinkComponent
N-[2-Methyl-3-(trifluoromethyl)phenyl]-N'-[2-methyl-6-(trifluoromethyl)pyridin-3-yl]urea
N-[2-Methyl-3-(trifluoromethyl)phenyl]-N'-[2-methyl-6-(trifluoromethyl)pyridin-3-yl]urea
Catalog # APOSPC300926-1G
Supplier:  Apollo Scientific
CAS Number:  
N-[2-Methyl-3-(trifluoromethyl)phenyl]-N'-[2-methyl-6-(trifluoromethyl)pyridin-3-yl]urea
Catalog # APOSPC300926-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :377.284 g/mol
Storage Temperature:Ambient
Formula:C₁₆H₁₃F₆N₃O
MDL Number:MFCD30481166