Order Entry
Netherlands
ContactUsLinkComponent
Methyl 2-{4-[2-methyl-3-(trifluoromethyl)phenyl]-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl}propanoate
Methyl 2-{4-[2-methyl-3-(trifluoromethyl)phenyl]-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl}propanoate
Catalog # APOSPC300922-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 2-{4-[2-methyl-3-(trifluoromethyl)phenyl]-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl}propanoate
Catalog # APOSPC300922-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :329.274 g/mol
Storage Temperature:Ambient
Formula:C₁₄H₁₄F₃N₃O₃
MDL Number:MFCD30481163