4-[6-(Trifluoromethyl)imidazo[1,2-a]pyridin-2-yl]benzonitrile
Catalog # APOSPC300864-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:287.239 g/mol
Storage Temperature:Ambient
CAS Number:907960-23-0
Formula:C₁₅H₈F₃N₃
MDL Number:MFCD28992256