Order Entry
Netherlands
ContactUsLinkComponent
1-[2,6-Dichloro-4-(trifluoromethyl)phenyl]-N-methyl-1H-pyrrole-2-sulphonamide
1-[2,6-Dichloro-4-(trifluoromethyl)phenyl]-N-methyl-1H-pyrrole-2-sulphonamide
Catalog # APOSPC300857-1G
Supplier:  Apollo Scientific
CAS Number:  
1-[2,6-Dichloro-4-(trifluoromethyl)phenyl]-N-methyl-1H-pyrrole-2-sulphonamide
Catalog # APOSPC300857-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :373.178 g/mol
Storage Temperature:Ambient
CAS Number:1858251-13-4
Formula:C₁₂H₉Cl₂F₃N₂O₂S
MDL Number:MFCD28954653