N-[2-Methyl-6-(trifluoromethyl)pyridin-3-yl]-N'-(tetrahydrofuran-2-ylmethyl)urea
Catalog # APOSPC300856-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:303.28 g/mol
Storage Temperature:Ambient
CAS Number:1858240-24-0
Formula:C₁₃H₁₆F₃N₃O₂
MDL Number:MFCD28954652