Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Nitro-1-phenoxy-2-(trifluoromethyl)benzene
4-Nitro-1-phenoxy-2-(trifluoromethyl)benzene
4-Nitro-1-phenoxy-2-(trifluoromethyl)benzene
4-Nitro-1-phenoxy-2-(trifluoromethyl)benzene
APOSPC300833-1G
:
Apollo Scientific
:
4-Nitro-1-phenoxy-2-(trifluoromethyl)benzene
APOSPC300833-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
283.203 g/mol
:
Ambient
:
6969-95-5
:
C₁₃H₈F₃NO₃
:
MFCD28347989