1-(4-Chlorophenoxy)-4-nitro-2-(trifluoromethyl)benzene
Catalog # APOSPC300832-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:317.648 g/mol
Storage Temperature:Ambient
CAS Number:21802-08-4
Formula:C₁₃H₇ClF₃NO₃
MDL Number:MFCD01579545