N-(4-Chlorophenyl)-N'-[3-(difluoromethoxy)phenyl]urea
Catalog # APOSPC300823-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:312.699 g/mol
Storage Temperature:Ambient
CAS Number:540771-25-3
Formula:C₁₄H₁₁ClF₂N₂O₂
MDL Number:MFCD28148243