
2,3,5-Trifluoro-4-methoxybenzonitrile
Catalog # APOSPC300803-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:187.119 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:916791-82-7
Formula:C₈H₄F₃NO
MDL Number:MFCD07782079