
6-(Trifluoromethyl)imidazo[1,2-a]pyridin-3-amine
Catalog # APOSPC300776-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:201.149 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1536950-06-7
Formula:C₈H₆F₃N₃
MDL Number:MFCD22076853