Order Entry
Netherlands
ContactUsLinkComponent
4-[2-Methyl-6-(trifluoromethyl)pyridin-3-yl]pyrimidine-2-thiol
4-[2-Methyl-6-(trifluoromethyl)pyridin-3-yl]pyrimidine-2-thiol
Catalog # APOSPC300772-1G
Supplier:  Apollo Scientific
CAS Number:  
4-[2-Methyl-6-(trifluoromethyl)pyridin-3-yl]pyrimidine-2-thiol
Catalog # APOSPC300772-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :271.262 g/mol
Storage Temperature:Ambient
CAS Number:1858250-86-8
Formula:C₁₁H₈F₃N₃S
MDL Number:MFCD27995628