Order Entry
Netherlands
ContactUsLinkComponent
Methyl 2,3,5-trifluoro-4-piperidin-1-ylbenzoate
Methyl 2,3,5-trifluoro-4-piperidin-1-ylbenzoate
Catalog # APOSPC300762-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 2,3,5-trifluoro-4-piperidin-1-ylbenzoate
Catalog # APOSPC300762-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :273.251 g/mol
Storage Temperature:Ambient
CAS Number:1858249-59-8
Formula:C₁₃H₁₄F₃NO₂
MDL Number:MFCD28040510