Order Entry
Netherlands
ContactUsLinkComponent
Ethyl 2,3,5-trifluoro-4-hydroxybenzoate
Irritant
Ethyl 2,3,5-trifluoro-4-hydroxybenzoate
Catalog # APOSPC300759-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Ethyl 2,3,5-trifluoro-4-hydroxybenzoate
Catalog # APOSPC300759-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :220.145 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1214351-42-4
Formula:C₉H₇F₃O₃
MDL Number:MFCD14698251