2,3,5-Trifluoro-4-hydrazinobenzhydrazide
Catalog # APOSPC300757-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:220.152 g/mol
Storage Temperature:Ambient
CAS Number:1823962-17-9
Formula:C₇H₇F₃N₄O
MDL Number:MFCD27995621