N-(2-Methoxyphenyl)-N'-[2-methyl-6-(trifluoromethyl)pyridin-3-yl]urea
Catalog # APOSPC300732-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:325.286 g/mol
Storage Temperature:Ambient
CAS Number:1227954-80-4
Formula:C₁₅H₁₄F₃N₃O₂
MDL Number:MFCD12406832