N-(2-Chlorophenyl)-N'-[2-methyl-6-(trifluoromethyl)pyridin-3-yl]urea
Catalog # APOSPC300730-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:329.705 g/mol
Storage Temperature:Ambient
Formula:C₁₄H₁₁ClF₃N₃O
MDL Number:MFCD12406830