N-Benzyl-N'-[2-methyl-6-(trifluoromethyl)pyridin-3-yl]urea
Catalog # APOSPC300660-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:309.286 g/mol
Storage Temperature:Ambient
CAS Number:1227954-87-1
Formula:C₁₅H₁₄F₃N₃O
MDL Number:MFCD12406821