4-(4-Bromophenoxy)-2-(trifluoromethyl)aniline
Catalog # APOSPC300625-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:332.116 g/mol
Storage Temperature:Ambient
CAS Number:1040341-48-7
Formula:C₁₃H₉BrF₃NO
MDL Number:MFCD12444002