4-(4-Bromophenoxy)-1-nitro-2-(trifluoromethyl)benzene
Catalog # APOSPC300624-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:362.099 g/mol
Storage Temperature:Ambient
CAS Number:1156314-30-5
Formula:C₁₃H₇BrF₃NO₃
MDL Number:MFCD13677626