
3-Bromo-N-methyl-5-(trifluoromethyl)pyridin-2-amine
Catalog # APOSPC300570-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:255.035 g/mol
:GHS07
Storage Temperature:Ambient
Formula:C₇H₆BrF₃N₂
MDL Number:MFCD16619584