
2-(1-Methylhydrazino)-6-(trifluoromethyl)pyridine
Catalog # APOSPC300569-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:191.154 g/mol
:GHS07
Storage Temperature:Ambient
Formula:C₇H₈F₃N₃
MDL Number:MFCD00052296