8-(Trifluoromethyl)imidazo[1,2-a]pyridine
Catalog # APOSPC300503-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:186.134 g/mol
Boiling Pt:110 °C (1 mm Hg)
Melting Pt:32…34 °C
Storage Temperature:Ambient
CAS Number:944580-98-7
Formula:C₈H₅F₃N₂
MDL Number:MFCD09994638