
(E)-1-(2,6-Difluorophenyl)-3-(dimethylamino)prop-2-en-1-one
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:138716-25-3
:
Formula:C₁₁H₁₁F₂NO
MDL Number:MFCD02089573
:211.208 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together