

3-Chloro-2-isothiocyanato-5-(trifluoromethyl)pyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1360887-00-8
:
Formula:C₇H₂ClF₃N₂S
MDL Number:MFCD22557535
:238.617 g/mol
UN:2923
ADR:8+6.1,III
:GHS05
:GHS06
Storage Temperature:Ambient
Frequently Bought Together