
3-Chloro-2-((difluoromethyl)sulphonyl)-5-(trifluoromethyl)pyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1823183-18-1
:
Formula:C₇H₃ClF₅NO₂S
MDL Number:MFCD29042616
:295.614 g/mol
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06
Frequently Bought Together