
1-Bromo-4-(difluoromethyl)-2-(trifluoromethyl)benzene 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1214358-07-2
:
Formula:C₈H₄BrF₅
MDL Number:MFCD14698541
:275.013 g/mol
Storage Temperature:Ambient
UN:3265
ADR:8,III
:GHS05
Frequently Bought Together