Ethyl 5-chloro-4-(trifluoromethyl)pyridine-2-carboxylate 97%
Catalog # APOSPC28225-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:253.606 g/mol
Storage Temperature:Ambient
CAS Number:1370587-22-6
Formula:C₉H₇ClF₃NO₂
MDL Number:MFCD22126112